Learn More
n-Hexatriacontane, 97+%
CAS: 630-06-8 | C36H74 | 506.99 g/mol
$108.40 - $328.34
Chemical Identifiers
| CAS | 630-06-8 |
|---|---|
| Molecular Formula | C36H74 |
| Molecular Weight (g/mol) | 506.99 |
| MDL Number | MFCD00009413 |
| InChI Key | YDLYQMBWCWFRAI-UHFFFAOYSA-N |
| Synonym | n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g |
| PubChem CID | 12412 |
| ChEBI | CHEBI:72688 |
| IUPAC Name | hexatriacontane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Description
n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 630-06-8 | |
| 506.99 | |
| YDLYQMBWCWFRAI-UHFFFAOYSA-N | |
| 12412 | |
| hexatriacontane |
| C36H74 | |
| MFCD00009413 | |
| n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g | |
| CHEBI:72688 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
| 630-06-8 | |
| White | |
| 497°C | |
| 1.4573 | |
| 10 g | |
| Insoluble in water. | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 506.99 | |
| CHEBI:72688 | |
| ≥97% | |
| n-Hexatriacontane |
| 76.2°C | |
| 0.779 g/mL | |
| C36H74 | |
| MFCD00009413 | |
| n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g | |
| YDLYQMBWCWFRAI-UHFFFAOYSA-N | |
| hexatriacontane | |
| 12412 | |
| 506.95 | |
| Wax |
Safety and Handling
EINECSNumber : 211-127-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only