missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 110-54-3 | C6H14 | 86.18 g/mol

Supplier:  Thermo Scientific Chemicals 0417279Z

 View more versions of this product

Catalog No. AA417279Z


Only null left
Explore available promotions

Description

Description

n-Hexane is used in the formulation of glues for shoes, leather products, and roofing, as a cleaning agent, de-greasing agent. It is used to extract edible oils from seeds and vegetables, a non-polar solvent in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

n-Hexane
110-54-3
69°C
-23°C (-9°F)
ChemSeal™ Bottle
1.375
MFCD02179311
1730733
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
Spectrophotometric
-95°C
0.659 g/mL
800 mL
Weak, characteristic
Liquid
C6H14
UN1208
14,4694
Soluble in ethyl ether, chloroform. Miscible with ethanol.
CCCCCC
86.18
CHEBI:29021
≥95%

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

EINECSNumber : 203-777-6

RTECSNumber : MN9275000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only