Learn More
n-Hexane, HPLC Grade, 95% min
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Supplier: Thermo Scientific Chemicals 039199K7
Description
as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Hexane | |
| 110-54-3 | |
| 69°C | |
| -23°C (-9°F) | |
| 1.375 | |
| C6H14 | |
| 1730733 | |
| Very soluble in ethanol,ethyl ether and chloroform. | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| HPLC |
| °C to 95°C | |
| 0.659 g/mL | |
| 4 L | |
| Liquid, Distilled In Glass | |
| 95% min | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 |
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 203-777-6
RTECSNumber : MN9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only