missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1043712500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1043712500 2.5 L Glass Bottle
M1043711000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1043712500 Supplier Technidata Supplier No. 1.04371.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -94.3°C
Color Colorless
Density 0.66g/cm3 at 20°C
Quantity 2.5 L
Flash Point −22°C (−7.6°F)
Ignition Point 240° (464°F)
Odor Benzine-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.37
Vapor Pressure 160 hPa (20°C)
UN Number 1208
Boiling Point 69°C
Solubility Information Soluble in water: 0.0095g/L at 20°C
Formula Weight 86.18
Chemical Name or Material Hexane
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H315: Causes skin irritation.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.