missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Environmental Grade, 95+%

Catalog No. AA42100K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42100K2 1 L
AA42100M1 4 x 1 L
AA42100K7 4 L
AA42100M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA42100K2 Supplier Thermo Scientific Chemicals Supplier No. 042100K2

For Pesticide Residue, Environmental and GC Analysis | CAS: 110-54-3 | C6H14 | 86.18 g/mol

Pesticide residue, environmental and GC analysis.n-Hexane is used in spectrophotometry and environmental testing. It is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point °C to 95°C
Color Colorless
Appearance Colorless liquid
Density 0.659
Quantity 1 L
Flash Point -23°C (-9F)
Ignition Point 240°C (464°F)
Odor Weak, characteristic
Physical Form Liquid
Refractive Index 1.375
Assay Percent Range >95%
Beilstein 1730733
Boiling Point 69°C
Solubility Information Very soluble in ethanol,ethyl ether and chloroform.
Formula Weight 86.18
Grade Environmental
Chemical Name or Material n-Hexane
Show More Show Less
Hazard Category H225-H304-H315-H319-H335-H336-H361f-H372
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-777-6
RTECSNumber MN9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only