missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, MilliporeSigma™

Catalog No. p-3176646
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX02956 1 L Glass Bottle
MHX02951 4 L Glass Bottle
MHX0295P1 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MHX02956 Supplier Technidata Supplier No. HX02956
Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Quantity 1 L
Packaging Glass Bottle
Assay Percent Range 95%
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
Chemical Name or Material n-Hexane, OmniPure™
Recommended Storage Room Temperature