Learn More
n-Hexane, anhydrous
Solvent in Sol-Gel Precursor Applications | CAS: 110-54-3 | C6H14 | 86.18 g/mol
$78.92 - $601.97
Chemical Identifiers
| CAS | 110-54-3 |
|---|---|
| Molecular Formula | C6H14 |
| Molecular Weight (g/mol) | 86.18 |
| MDL Number | MFCD02179311 |
| InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
| Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
| PubChem CID | 8058 |
| ChEBI | CHEBI:29021 |
| IUPAC Name | hexane |
| SMILES | CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA43263AK
|
Thermo Scientific Chemicals
043263AK |
250 mL |
Each for $78.92
|
|
|||||
|
AA43263K2
|
Thermo Scientific Chemicals
043263K2 |
1 L |
Each for $180.05
|
|
|||||
|
AA43263K7
|
Thermo Scientific Chemicals
043263K7 |
4 L |
Each for $601.97
|
|
|||||
Description
n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Specifications
| °C to 95°C | |
| Colorless | |
| 69°C | |
| -23°C (-9°F) | |
| Weak, characteristic | |
| 1.375 | |
| MFCD02179311 | |
| 1208 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 |
| 110-54-3 | |
| 0.659 g/cm3 at 20°C | |
| 250 mL | |
| 240°C (464°F) | |
| Liquid | |
| C6H14 | |
| 120mm Hg at 20°C | |
| 1730733 | |
| Miscible with alcohol,chloroform,acetone and ether. | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| n-Hexane |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 203-777-6
RTECSNumber : MN9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only