missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 99%

CAS: 110-54-3 | C6H14 | 86.18 g/mol

Supplier:  Thermo Scientific Chemicals L09938AU

 View more versions of this product

Catalog No. AAL09938AU

Only null left
Explore available promotions

Description

Description

n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.

Solubility
Miscible with alcohol, chloroform, acetone and ether.

Notes
Incompatible with dinitrogen tetraoxide. Store in a cool and dry environment.
Specifications

Specifications

n-Hexane
110-54-3
69°C
−23°C (−9°F)
1.375
MFCD02179311
1730733
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
-95°C
0.659
1000 mL
Weak, Characteristic
C6H14
UN1208
14,4694
Miscible with alcohol,chloroform,acetone and ether.
CCCCCC
86.18
CHEBI:29021
99%

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

EINECSNumber : 203-777-6

RTECSNumber : MN9275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only