Learn More
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
CAS: 110-54-3 | C6H14 | 86.18 g/mol
$170.05 - $170.05
Chemical Identifiers
| CAS | 110-54-3 |
|---|---|
| Molecular Formula | C6H14 |
| Molecular Weight (g/mol) | 86.18 |
| MDL Number | MFCD02179311 |
| InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
| Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
| PubChem CID | 8058 |
| ChEBI | CHEBI:29021 |
| IUPAC Name | hexane |
| SMILES | CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC326780010
|
Thermo Scientific Chemicals
326780010 |
1 L | Glass Bottle |
Each for $170.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysisChemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Specifications
| -95.0°C | |
| 0.004 max. at 270nm, 0.097 max. at 220nm, 0.602 max. at 200nm | |
| 100.0 | |
| 69.0°C | |
| −22°C | |
| Liquid | |
| 0.0002% max. | |
| 0.659 | |
| CH3(CH2)4CH3 | |
| 0.31 mPa.s (20°C) | |
| 01, 142 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 | |
| Residue Analysis |
| 110-54-3 | |
| 94.0 | |
| 0.6590g/mL | |
| 1 L | |
| Glass Bottle | |
| 1.3748 to 1.3810 | |
| 95% | |
| C6H14 | |
| MFCD02179311 | |
| 0.005% max. (Coulometric) | |
| 15, 4732 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| 95% | |
| n-Hexane |
Safety and Handling
GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.
Danger
EINECSNumber : 203-777-6
RTECSNumber : MN9275000
TSCA : TSCA
RUO – Research Use Only