missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane 95%, HPLC, MilliporeSigma™

Catalog No. MHX02911
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX02911 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX02911 Supplier Technidata Supplier No. HX02911

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets Reagent Specifications for testing USP/NF monographs

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -139.4°C
Color Colorless
Density 0.66g/cm3 at 20°C
Quantity 4 L
Flash Point −23.15°C (−9.7°F), Closed cup
Ignition Point 224.85°C (436.7°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
UN Number 1208
Solubility Information Partially soluble in water
Boiling Point 69°C
Formula Weight 86.18
Percent Purity ≥99.0%
Grade HPLC
Chemical Name or Material n-Hexanes
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept uprightto prevent leakage.