missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 95+%, ACS reagent

Catalog No. AC445780010
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445780010 1 L
AC445780025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445780010 Supplier Thermo Scientific Chemicals Supplier No. 445780010

missing translation for 'validMainframeMsg'

CAS: 110-54-3 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95.0°C
Density 0.6590 g/mL
Quantity 1 L
Flash Point -22°C
Refractive Index 1.3748 to 1.3810
Specific Gravity 0.659
Assay Percent Range 95+%
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
Merck Index 15, 4732
Boiling Point 69.0°C
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Grade ACS
Chemical Name or Material n-Hexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May be fatal if swallowed and enters airways.

May cause drowsiness or dizziness.

Causes skin irritation.

May cause damage to organs through prolonged or repeated exposure.

Toxic to aquatic life with long las
Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Use personal protective equipment as required.

Do not breathe dust/fume/gas/mist/vapors/spray.

IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

Do NOT induce vomiting.

Avoid release to the environment.

IF ON SKIN: Wash with plenty of soap and water.

EINECSNumber 203-777-6

RUO – Research Use Only