missing translation for 'onlineSavingsMsg'
Learn More

(n-Hexadecyl)tri-n-butylphosphonium bromide, 98+%

Catalog No. AAL0133514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0133514 25 g
AAL0133522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0133514 Supplier Thermo Scientific Chemicals Supplier No. L0133514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14937-45-2 | C28H60BrP | 507.67 g/mol

(n-Hexadecyl)tri-n-butylphosphonium bromide is used as Phase-transfer-catalyst. It is a eco friendly highly effective water treatment agent with low foaming and toxic-free.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(n-Hexadecyl)tri-n-butylphosphonium bromide is used as Phase-transfer-catalyst. It is a eco friendly highly effective water treatment agent with low foaming and toxic-free.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Protect from carbondioxide. Store away from moisture.

Chemical Identifiers

CAS 14937-45-2
Molecular Formula C28H60BrP
Molecular Weight (g/mol) 507.67
MDL Number MFCD00011775
InChI Key RYVBINGWVJJDPU-UHFFFAOYSA-M
Synonym tributylhexadecylphosphonium bromide, tributyl hexadecyl phosphonium bromide, cetyltributylphosphonium bromide, tributyl hexadecyl phosphanium bromide, phosphonium, tributylhexadecyl-, bromide, hexadecyltributylphosphonium bromide, phosphonium, tributylhexadecyl-, bromide 1:1, hexadecyltri-n-butylphosphonium bromide, n-hexadecyl tri-n-butylphosphonium bromide, tbhdpb
PubChem CID 84716
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 56°C to 62°C
Quantity 25 g
Beilstein 4167404
Sensitivity Hygroscopic
Solubility Information Soluble in methanol.
IUPAC Name tributyl(hexadecyl)phosphanium bromide
Formula Weight 507.67
Percent Purity ≥98%
Chemical Name or Material (n-Hexadecyl)tri-n-butylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-014-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only