missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl palmitate, 98%

CAS: 540-10-3 | C32H64O2 | 480.86 g/mol

Chemical Identifiers

CAS 540-10-3
Molecular Formula C32H64O2
Molecular Weight (g/mol) 480.86
MDL Number MFCD00053739
InChI Key PXDJXZJSCPSGGI-UHFFFAOYSA-N
Synonym cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PubChem CID 10889
ChEBI CHEBI:75584
IUPAC Name hexadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC338460050
View Documents Promotion Details
N/A 5 g Glass bottle N/A
Only null left
 
AC338460250
View Documents Promotion Details
N/A 25 g Glass bottle N/A
Only null left
 
AC338461000
View Documents Promotion Details
N/A 100 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

540-10-3
480.86
PXDJXZJSCPSGGI-UHFFFAOYSA-N
10889
hexadecyl hexadecanoate
C32H64O2
MFCD00053739
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
CHEBI:75584
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

540-10-3
100.0
White
Authentic
C32H64O2
MFCD00053739
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PXDJXZJSCPSGGI-UHFFFAOYSA-N
hexadecyl hexadecanoate
10889
480.86
Crystalline Powder
97.5
52°C to 56°C
360°C
98%
CH3(CH2)14COO(CH2)15CH3
15, 2030
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
480.86
CHEBI:75584
98%
n-Hexadecyl palmitate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-736-6

SDS
Documents

Documents

RUO – Research Use Only