missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, 99%, anhydrous, AcroSeal™

CAS: 544-76-3 | C16H34 | 226.44 g/mol

$531.75 - $531.75

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.44
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436140010
SDS View Documents
Thermo Scientific Chemicals
436140010
1 L AcroSeal™ Glass Bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

544-76-3
226.44
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CHEBI:45296
CCCCCCCCCCCCCCCC
C16H34
DCAYPVUWAIABOU-UHFFFAOYSA-N
11006
hexadecane

Specifications

544-76-3
100.0
Colorless
287°C
Authentic
AcroSeal™ Glass Bottle
1.4330 to 1.435
1 L
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
DCAYPVUWAIABOU-UHFFFAOYSA-N
hexadecane
11006
CHEBI:45296
99%
n-Hexadecane
98.5
18°C
0.7730g/mL
135°C
98.5% min. (GC)
C16H34
CH3(CH2)14CH3
0.773
Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone and ether
CCCCCCCCCCCCCCCC
226.44
3.032 mPa.s (25°C)
226.44
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-878-9

SDS
Documents

Documents

RUO – Research Use Only