missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, 95%

Catalog No. p-7058805
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43283LV 1 gal.
AA43283LW 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43283LV Supplier Thermo Scientific Chemicals Supplier No. 043283LV
Only null left

Standard for rating diesel-fuel ignition qualities | CAS: 544-76-3 | C16H34 | 226.448 g/mol

Standard for rating diesel-fuel ignition qualitiesHexadecane serves as a reference for other fuel mixtures and is assigned cetane number of 100; used as shorthand for cetane number, an indicator of the combustion speed of diesel fuel. Hexadecane is used in the preparation of emulsions; also used as a substrate for the production of biosurfactants. n-Hexadecane has been found useful in the identification of the wheat seed protein CM3 as a highly active emulsifier. Superamphiphobicity on polymers can be studied with hexadecane immersion tests. Shape-stabilized phase change material (SSPCM), prepared using hexadecane as a PCM, into xGnP as a supporting material, has been demonstrated to have the potential to be used in developing building materials with improved thermal energy storage charecteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Melting Point 18°C
Color Colorless
Density 0.773 g/mL
Boiling Point 287°C
Flash Point 135°C (275°F)
Refractive Index 1.4345
Quantity 1 gal.
Beilstein 1736592
Solubility Information Soluble in ethanolMiscible with alcohol,acetone,chloroform and ether; Immiscible with water.
Formula Weight 226.45
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material n-Hexadecane
Show More Show Less
Hazard Category H304
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 208-878-9
RTECSNumber ML9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only