missing translation for 'onlineSavingsMsg'
Learn More

n-Heptacosane, 97%

Catalog No. p-7044689
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL07796MD 250 mg
AAL0779603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL07796MD Supplier Thermo Scientific Chemicals Supplier No. L07796MD
Only null left

CAS: 593-49-7 | C27H56 | 380.745 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 593-49-7
Molecular Formula C27H56
Molecular Weight (g/mol) 380.745
MDL Number MFCD00009862
InChI Key BJQWYEJQWHSSCJ-UHFFFAOYSA-N
Synonym n-heptacosane, unii-vp371w2gjs, vp371w2gjs, hexacosane, methyl, acmc-1aog8, qspl 047, qspl 073, heptacosane, analytical standard, ch3-ch2 25-ch3, heptacosane gc
PubChem CID 11636
ChEBI CHEBI:32941
IUPAC Name heptacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 57°C to 60°C
Boiling Point 270°C (15 mmHg)
Flash Point 120°C (248°F)
Quantity 250 mg
Beilstein 1706242
Formula Weight 380.74
Percent Purity 97%
Chemical Name or Material n-Heptacosane
EINECSNumber 209-792-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only