missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heneicosane, 99%
CAS: 629-94-7 | C21H44 | 296.583 g/mol
Supplier: Thermo Scientific Chemicals A1819806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Heneicosane | |
| 39°C to 43°C | |
| 100°C (2 mmHg) | |
| C21H44 | |
| 5 g | |
| heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj | |
| CCCCCCCCCCCCCCCCCCCCC | |
| 296.583 | |
| CHEBI:32931 | |
| 99% |
| 629-94-7 | |
| 0.792 | |
| >110°C (230°F) | |
| MFCD00009346 | |
| 1748500 | |
| FNAZRRHPUDJQCJ-UHFFFAOYSA-N | |
| henicosane | |
| 12403 | |
| 296.58 |
Chemical Identifiers
| 629-94-7 | |
| 296.583 | |
| FNAZRRHPUDJQCJ-UHFFFAOYSA-N | |
| 12403 | |
| henicosane |
| C21H44 | |
| MFCD00009346 | |
| heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj | |
| CHEBI:32931 | |
| CCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : 211-118-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only