missing translation for 'onlineSavingsMsg'
Learn More

n-Heneicosane, 99%

Catalog No. AAA1819803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1819803 1 g
AAA1819806 5 g
AAA1819814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1819803 Supplier Thermo Scientific Chemicals Supplier No. A1819803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-94-7 | C21H44 | 296.583 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-94-7
Molecular Formula C21H44
Molecular Weight (g/mol) 296.583
MDL Number MFCD00009346
InChI Key FNAZRRHPUDJQCJ-UHFFFAOYSA-N
Synonym heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
PubChem CID 12403
ChEBI CHEBI:32931
IUPAC Name henicosane
SMILES CCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 39°C to 43°C
Density 0.792
Boiling Point 100°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 1748500
Formula Weight 296.58
Percent Purity 99%
Chemical Name or Material n-Heneicosane
EINECSNumber 211-118-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only