missing translation for 'onlineSavingsMsg'
Learn More

n-Heneicosane, 98%

CAS: 629-94-7 | C21H44 | 296.58 g/mol

$1,290.74 - $1,290.74

Chemical Identifiers

CAS 629-94-7
Molecular Formula C21H44
Molecular Weight (g/mol) 296.58
MDL Number MFCD00009346
InChI Key FNAZRRHPUDJQCJ-UHFFFAOYSA-N
Synonym heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
PubChem CID 12403
ChEBI CHEBI:32931
IUPAC Name henicosane
SMILES CCCCCCCCCCCCCCCCCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC352770500
SDS View Documents
Thermo Scientific Chemicals
352770500
50 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-94-7
296.58
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
12403
henicosane
C21H44
MFCD00009346
heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
CHEBI:32931
CCCCCCCCCCCCCCCCCCCCC

Specifications

629-94-7
100.0
White
>110°C
97.5% min. (GC)
C21H44
MFCD00009346
heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
henicosane
12403
296.58
Crystalline Chunks, Crystals, or Flakes
97.5
39°C to 43°C
100°C (2.0 mmHg)
Authentic
Glass bottle
CH3(CH2)19CH3
50 g
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether
CCCCCCCCCCCCCCCCCCCCC
296.58
CHEBI:32931
98%
n-Heneicosane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-118-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only