Learn More
N-Guanylurea sulfate, 97%
CAS: 591-01-5 | C4H12N8O2·H2SO4
Supplier: Thermo Scientific Chemicals A1910618
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Guanylurea sulfate | |
| ∼195°C (decomposition) | |
| MFCD00035565 | |
| 6099371 | |
| Dicyanodiamidine sulfate | |
| 97% |
| 591-01-5 | |
| C4H12N8O2·H2SO4 | |
| 50 g | |
| 14,3093 | |
| 302.27 |
Chemical Identifiers
| 591-01-5 | |
| C4H12N8O2·H2SO4 | |
| MFCD00035565 | |
| Dicyanodiamidine sulfate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 209-697-8
TSCA : Yes
RUO – Research Use Only