missing translation for 'onlineSavingsMsg'
Learn More

N-Guanylurea sulfate, 97%

Catalog No. AAA1910630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1910630 250 g
AAA1910609 10 g
AAA1910618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1910630 Supplier Thermo Scientific Chemicals Supplier No. A1910630

CAS: 591-01-5 | C4H12N8O2·H2SO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-01-5
Molecular Formula C4H12N8O2·H2SO4
MDL Number MFCD00035565
Synonym Dicyanodiamidine sulfate

Specifications

Melting Point ∼195°C (decomposition)
Quantity 250 g
Beilstein 6099371
Merck Index 14,3093
Formula Weight 302.27
Percent Purity 97%
Chemical Name or Material N-Guanylurea sulfate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-697-8
TSCA Yes

RUO – Research Use Only