missing translation for 'onlineSavingsMsg'
Learn More

N-Glycyl-L-tyrosine, 98%

Catalog No. p-7056460
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6538806 5 g
AAH6538822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6538806 Supplier Thermo Scientific Chemicals Supplier No. H6538806
Only null left

CAS: 658-79-7 | C11H14N2O4 | 238.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 658-79-7
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.24
MDL Number MFCD00008126
InChI Key XBGGUPMXALFZOT-IMWMWJONNA-N
Synonym glycyl-l-tyrosine, gly-tyr, n-glycyl-l-tyrosine, h-gly-tyr-oh, glycyltyrosine, l-tyrosine, glycyl, s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, chembl53400, 2s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2-aminoacetyl amino-3-4-hydroxyphenyl propanoic acid
PubChem CID 92829
ChEBI CHEBI:73517
IUPAC Name (2S)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoic acid
SMILES NCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White to Yellow
Beilstein 2700715
Quantity 5 g
Formula Weight 238.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Glycyl-L-tyrosine
EINECSNumber 211-525-1
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only