missing translation for 'onlineSavingsMsg'
Learn More

N-Glycolylneuraminic Acid 98.0+%, TCI America™

Catalog No. g033610mg
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G033610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G033610MG Supplier TCI America Supplier No. G033610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1113-83-3
Molecular Formula C11H19NO10
Molecular Weight (g/mol) 325.27
MDL Number MFCD00057551
InChI Key FDJKUWYYUZCUJX-GETJOPHTSA-N
Synonym Neu5Gc
PubChem CID 71317108
IUPAC Name (2S,4S,5R,6R)-2,4-dihydroxy-5-[(2-hydroxyacetyl)amino]-6-[(2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES C1C(C(C(OC1(C(=O)O)O)C(C(CO)O)O)NC(=O)CO)O

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 325.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Glycolylneuraminic Acid
TSCA Yes
Recommended Storage Freezer