missing translation for 'onlineSavingsMsg'
Learn More

N-Formylethylamine 98.0+%, TCI America™

Catalog No. F0188500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0188500ML 500 mL
F018825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0188500ML Supplier TCI America Supplier No. F0188500ML
Only null left

Chemical Identifiers

CAS 627-45-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.095
MDL Number MFCD00042672
InChI Key KERBAAIBDHEFDD-UHFFFAOYSA-N
Synonym N-Ethylformamide
PubChem CID 12319
IUPAC Name N-ethylformamide
SMILES CCNC=O

Specifications

Color Yellow
Boiling Point 202°C
Quantity 500 mL
Formula Weight 73.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Formylethylamine
RTECSNumber LQ2725000
TSCA Yes