missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Formylethylamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 202°C |
| Quantity | 25 mL |
| Formula Weight | 73.10 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | N-Formylethylamine |
Safety and Handling
| RTECSNumber | LQ2725000 |
| TSCA | Yes |