missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-proline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 456420050

 View more versions of this product

Catalog No. AC456420050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-Formyl-L-proline
Authentic
5g
1-formyl-l-proline, l-proline, 1-formyl, n-formylproline, n-formyl-l-proline, 2s-1-formylpyrrolidine-2-carboxylic acid, s---n-formylproline, n-formyl-l-proline tlc, s-1-formyl-2-pyrrolidinecarboxylic acid, s-1-formylpyrrolidine-2-carboxylic acid
C1CC(N(C1)C=O)C(=O)O
143.14
143.14
13200-83-4
C6H9NO3
-125
DHDRGOURKDLAOT-YFKPBYRVSA-N
(2S)-1-formylpyrrolidine-2-carboxylic acid
12461843
97%

Chemical Identifiers

13200-83-4
143.14
1-formyl-l-proline, l-proline, 1-formyl, n-formylproline, n-formyl-l-proline, 2s-1-formylpyrrolidine-2-carboxylic acid, s---n-formylproline, n-formyl-l-proline tlc, s-1-formyl-2-pyrrolidinecarboxylic acid, s-1-formylpyrrolidine-2-carboxylic acid
(2S)-1-formylpyrrolidine-2-carboxylic acid
C6H9NO3
DHDRGOURKDLAOT-YFKPBYRVSA-N
12461843
C1CC(N(C1)C=O)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 236-165-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only