missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%, Thermo Scientific Chemicals

Catalog No. p-7055115
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6361406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6361406 Supplier Thermo Scientific Chemicals Supplier No. H6361406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 197632-77-2 | Molecular Formula: C23H27NO5 | Molecular Weight: 397.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 197632-77-2
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
MDL Number MFCD02094430
InChI Key PQSAXALAXPNFMG-FQEVSTJZSA-N
Synonym fmoc-n-me-ser tbu-oh, n-fmoc-n-methyl-o-tert-butyl-l-serine, fmoc-n-methyl-o-t-butyl-l-serine, fmoc-n-methyl-o-tert-butyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-serine, 2s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy propanoic acid, fmoc-meser tbu-oh, pubchem19046, fmoc-n-me-serine tbu-oh
PubChem CID 7010370
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 213°C to 218°C
Color White
Quantity 5 g
Formula Weight 397.46
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N-Fmoc-O-tert-butyl-N-methyl-L-serine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only