Learn More
N-Fmoc-O-tert-butyl-D-tyrosine, 95%
CAS: 118488-18-9 | C28H29NO5 | 459.54 g/mol
Supplier: Thermo Scientific Chemicals H6276506
Description
N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
Solubility
Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
| N-Fmoc-O-tert-butyl-D-tyrosine | |
| C28H29NO5 | |
| 5 g | |
| fmoc-d-tyr tbu-oh, fmoc-o-tert-butyl-d-tyrosine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, nalpha-9h-fluoren-9-ylmethoxy carbonyl-o-tert-butyl-d-tyrosine, 2r-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-tyrosine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, ambotzfaa1341, pubchem10054, fmoc-d-tyr but-oh, fmoc-o-tert.butyl-d-tyrosine | |
| JAUKCFULLJFBFN-KSYWNVGFNA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid | |
| 12968135 | |
| 95% |
| 118488-18-9 | |
| MFCD00065684 | |
| 6691868 | |
| Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water. | |
| CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1 | |
| 459.54 | |
| 459.53 |
Chemical Identifiers
| 118488-18-9 | |
| 459.54 | |
| JAUKCFULLJFBFN-KSYWNVGFNA-N | |
| 12968135 | |
| CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1 |
| C28H29NO5 | |
| MFCD00065684 | |
| fmoc-d-tyr tbu-oh, fmoc-o-tert-butyl-d-tyrosine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, nalpha-9h-fluoren-9-ylmethoxy carbonyl-o-tert-butyl-d-tyrosine, 2r-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-tyrosine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, ambotzfaa1341, pubchem10054, fmoc-d-tyr but-oh, fmoc-o-tert.butyl-d-tyrosine | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only