missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-methyl-D-tyrosine, 98%

CAS: 201335-88-8 | C25H23NO5 | 417.46 g/mol

$96.56 - $323.05

Chemical Identifiers

CAS 201335-88-8
Molecular Formula C25H23NO5
Molecular Weight (g/mol) 417.46
MDL Number MFCD00237393
InChI Key JYQODLWFOPCSCS-UHFFFAOYNA-N
Synonym fmoc-d-tyr me-oh, fmoc-o-methyl-d-tyrosine, fmoc-d-4-methoxyphenylalanine, fmoc-4-methoxy-d-phenylalanine, fmoc-4-methoxy-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, fmoc-d-4-methoxy-phe-oh, fmoc-d-tyr me, fmoc-d-4-meo-phe-oh
PubChem CID 7019819
SMILES COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6307303
View Documents
Thermo Scientific Chemicals
H6307303
1 g
Each for $96.56
Only null left
 
AAH6307306
View Documents
Thermo Scientific Chemicals
H6307306
5 g
Each for $323.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

201335-88-8
417.46
JYQODLWFOPCSCS-UHFFFAOYNA-N
7019819
C25H23NO5
MFCD00237393
fmoc-d-tyr me-oh, fmoc-o-methyl-d-tyrosine, fmoc-d-4-methoxyphenylalanine, fmoc-4-methoxy-d-phenylalanine, fmoc-4-methoxy-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, fmoc-d-4-methoxy-phe-oh, fmoc-d-tyr me, fmoc-d-4-meo-phe-oh
COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1

Specifications

201335-88-8
C25H23NO5
fmoc-d-tyr me-oh, fmoc-o-methyl-d-tyrosine, fmoc-d-4-methoxyphenylalanine, fmoc-4-methoxy-d-phenylalanine, fmoc-4-methoxy-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, fmoc-d-4-methoxy-phe-oh, fmoc-d-tyr me, fmoc-d-4-meo-phe-oh
COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
1 g
7019819
98%
N-Fmoc-O-methyl-D-tyrosine
White
MFCD00237393
JYQODLWFOPCSCS-UHFFFAOYNA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methoxyphenyl)propanoic acid
417.46
417.45
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only