missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-benzylphospho-L-threonine, 97%

CAS: 175291-56-2 | C26H26NO8P | 511.47 g/mol

Supplier:  Thermo Scientific Chemicals H6300603

 View more versions of this product

Catalog No. AAH6300603

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Fmoc-O-benzylphospho-L-threonine
C26H26NO8P
7557104
HOFDVXHILSPFNS-OSPHWJPCSA-N
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid
511.47
511.46
97%
175291-56-2
MFCD00797870
fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh
C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1 g
15224585
Moisture sensitive
Solid

Chemical Identifiers

175291-56-2
511.47
HOFDVXHILSPFNS-OSPHWJPCSA-N
15224585
C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
C26H26NO8P
MFCD00797870
fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only