missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-benzylphospho-L-threonine, 97%

Catalog No. AAH63006MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63006MD 250 mg
AAH6300603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63006MD Supplier Thermo Scientific Chemicals Supplier No. H63006MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 175291-56-2 | C26H26NO8P | 511.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175291-56-2
Molecular Formula C26H26NO8P
Molecular Weight (g/mol) 511.47
MDL Number MFCD00797870
InChI Key HOFDVXHILSPFNS-OSPHWJPCSA-N
Synonym fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh
PubChem CID 15224585
IUPAC Name (2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid
SMILES C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Beilstein 7557104
Quantity 250 mg
Formula Weight 511.46
Sensitivity Moisture sensitive
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Fmoc-O-benzylphospho-L-threonine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only