missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-O-benzyl-L-serine, 95%
CAS: 83792-48-7 | C25H23NO5 | 417.461 g/mol
$175.81 - $594.90
Chemical Identifiers
| CAS | 83792-48-7 |
|---|---|
| Molecular Formula | C25H23NO5 |
| Molecular Weight (g/mol) | 417.461 |
| MDL Number | MFCD00065674 |
| InChI Key | DYBDGLCDMLNEMJ-QHCPKHFHSA-N |
| Synonym | fmoc-ser bzl-oh, fmoc-o-benzyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-l-serine, l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl, 2s-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmocser bzl, fmoc-serine bzl-oh, fmoc-l-ser bzl-oh, fmoc-s-2-amino-3-benzyloxypropionic acid, fmoc-ser o-bzl |
| PubChem CID | 7017914 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxypropanoic acid |
| SMILES | C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6658606
|
Thermo Scientific Chemicals
H6658606 |
5 g |
Each for $175.81
|
|
|||||
|
AAH6658614
|
Thermo Scientific Chemicals
H6658614 |
25 g |
Each for $594.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83792-48-7 | |
| 417.461 | |
| DYBDGLCDMLNEMJ-QHCPKHFHSA-N | |
| 7017914 | |
| C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
| C25H23NO5 | |
| MFCD00065674 | |
| fmoc-ser bzl-oh, fmoc-o-benzyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-l-serine, l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl, 2s-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmocser bzl, fmoc-serine bzl-oh, fmoc-l-ser bzl-oh, fmoc-s-2-amino-3-benzyloxypropionic acid, fmoc-ser o-bzl | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxypropanoic acid |
Specifications
| 83792-48-7 | |
| C25H23NO5 | |
| 3573590 | |
| DYBDGLCDMLNEMJ-QHCPKHFHSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxypropanoic acid | |
| 417.461 | |
| 417.46 | |
| Powder |
| Brown | |
| MFCD00065674 | |
| fmoc-ser bzl-oh, fmoc-o-benzyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-l-serine, l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl, 2s-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmocser bzl, fmoc-serine bzl-oh, fmoc-l-ser bzl-oh, fmoc-s-2-amino-3-benzyloxypropionic acid, fmoc-ser o-bzl | |
| C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
| 5 g | |
| 7017914 | |
| 95% | |
| N-Fmoc-O-benzyl-L-serine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only