missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-benzyl-L-serine, 95%

Catalog No. AAH6658606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6658606 5 g
AAH6658614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6658606 Supplier Thermo Scientific Chemicals Supplier No. H6658606

CAS: 83792-48-7 | C25H23NO5 | 417.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83792-48-7
Molecular Formula C25H23NO5
Molecular Weight (g/mol) 417.461
MDL Number MFCD00065674
InChI Key DYBDGLCDMLNEMJ-QHCPKHFHSA-N
Synonym fmoc-ser bzl-oh, fmoc-o-benzyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-l-serine, l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl, 2s-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmocser bzl, fmoc-serine bzl-oh, fmoc-l-ser bzl-oh, fmoc-s-2-amino-3-benzyloxypropionic acid, fmoc-ser o-bzl
PubChem CID 7017914
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color Brown
Beilstein 3573590
Quantity 5 g
Formula Weight 417.46
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-O-benzyl-L-serine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only