missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N-α-FMOC-N-Trityl-L-histidine, 98%

$61.66 - $143.37

Chemical Identifiers

CAS 109425-51-6
Molecular Formula C40H33N3O4
Molecular Weight (g/mol) 619.72
InChI Key XXMYDXUIZKNHDT-QNGWXLTQSA-N
Synonym fmoc-his trt-oh, n-fmoc-n'-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, a-fmoc-n im-trityl-l-histidine, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, ambotzfaa1090
PubChem CID 11422193
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC360330050
View Documents Promotion Details
Thermo Scientific™
360330050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC360330250
View Documents Promotion Details
Thermo Scientific™
360330250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

109425-51-6
619.72
fmoc-his trt-oh, n-fmoc-n'-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, a-fmoc-n im-trityl-l-histidine, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, ambotzfaa1090
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
C40H33N3O4
XXMYDXUIZKNHDT-QNGWXLTQSA-N
11422193
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

109425-51-6
100.0
97.5% min. (HPLC)
C40H33N3O4
fmoc-his trt-oh, n-fmoc-n'-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, a-fmoc-n im-trityl-l-histidine, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, ambotzfaa1090
5% max. 2-propanol, THF
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
619.72
619.72
N-α-FMOC-N-Trityl-L-histidine
97.5
Authentic
Glass bottle
83.3
Solubility in water: insoluble. Other solubilities: soluble in chloroform
XXMYDXUIZKNHDT-QNGWXLTQSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
11422193
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only