missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-N-methyl-L-valine, 95%
CAS: 84000-11-3 | C21H23NO4 | 353.42 g/mol
$165.23 - $626.74
Chemical Identifiers
| CAS | 84000-11-3 |
|---|---|
| Molecular Formula | C21H23NO4 |
| Molecular Weight (g/mol) | 353.42 |
| MDL Number | MFCD00153395 |
| InChI Key | YCXXXPZNQXXRIG-IBGZPJMESA-N |
| Synonym | fmoc-n-me-val-oh, fmoc-n-methyl-l-valine, n-fmoc-n-methyl-l-valine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl-l-valine, l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-3-methyl-butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1411, fmoc-n-me-l-val-oh |
| PubChem CID | 978328 |
| IUPAC Name | (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid |
| SMILES | CC(C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6367103
|
Thermo Scientific Chemicals
H6367103 |
1 g |
Each for $165.23
|
|
|||||
|
AAH6367106
|
Thermo Scientific Chemicals
H6367106 |
5 g |
Each for $626.74
|
|
|||||
Description
N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.
Solubility
Slightly soluble in water and dimethyl formamide.
Notes
Incompatible with strong oxidizing agents, reducing agents, bases and alkali metals. Store in a cool place.
Chemical Identifiers
| 84000-11-3 | |
| 353.42 | |
| YCXXXPZNQXXRIG-IBGZPJMESA-N | |
| 978328 | |
| CC(C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C21H23NO4 | |
| MFCD00153395 | |
| fmoc-n-me-val-oh, fmoc-n-methyl-l-valine, n-fmoc-n-methyl-l-valine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl-l-valine, l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-3-methyl-butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1411, fmoc-n-me-l-val-oh | |
| (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid |
Specifications
| 187°C to 190°C | |
| White | |
| MFCD00153395 | |
| Slightly soluble in water and dimethyl formamide. | |
| CC(C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 1 g | |
| 978328 | |
| 95% | |
| N-Fmoc-N-methyl-L-valine |
| 84000-11-3 | |
| C21H23NO4 | |
| fmoc-n-me-val-oh, fmoc-n-methyl-l-valine, n-fmoc-n-methyl-l-valine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl-l-valine, l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-3-methyl-butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1411, fmoc-n-me-l-val-oh | |
| YCXXXPZNQXXRIG-IBGZPJMESA-N | |
| (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid | |
| 353.42 | |
| 353.41 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only