missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-leucine, 98%

Catalog No. AAH63453MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63453MD 250 mg
AAH6345303 1 g
AAH6345306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63453MD Supplier Thermo Scientific Chemicals Supplier No. H63453MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103478-62-2 | C22H25NO4 | 367.45 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 103478-62-2
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00151933
InChI Key BUJQSIPFDWLNDC-GNLPSFAGNA-N
Synonym fmoc-n-methyl-l-leucine, fmoc-n-me-leu-oh, n-fmoc-n-methyl-l-leucine, fmoc-meleu-oh, l-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, pubchem13155, fmoc-n-a-methyl-l-leucine, n-methyl-n-fmoc-l-leu-oh
PubChem CID 7015835
SMILES CC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 113°C to 116°C
Color White
Beilstein 3630430
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
Quantity 250 mg
Formula Weight 367.44
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-N-methyl-L-leucine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only