missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-isoleucine, 95%, Thermo Scientific Chemicals

$522.53 - $522.53

Chemical Identifiers

CAS 138775-22-1
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00153389
InChI Key IQIOLCJHRZWOLS-DZLYFNQKNA-N
Synonym fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
PubChem CID 15433034
IUPAC Name (2S,3S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6364006
View Documents
Thermo Scientific Chemicals
H6364006
5 g
Each for $522.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

138775-22-1
367.45
IQIOLCJHRZWOLS-DZLYFNQKNA-N
15433034
CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C22H25NO4
MFCD00153389
fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
(2S,3S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylpentanoic acid

Specifications

138775-22-1
MFCD00153389
fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
5 g
15433034
95%
N-Fmoc-N-methyl-L-isoleucine
C22H25NO4
7548444
IQIOLCJHRZWOLS-DZLYFNQKNA-N
(2S,3S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylpentanoic acid
367.45
367.44
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only