missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-D-leucine, 97%

Catalog No. AAH6389503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6389503 1 g
AAH6389506 5 g
AAH6389514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6389503 Supplier Thermo Scientific Chemicals Supplier No. H6389503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103478-63-3 | C22H25NO4 | 367.45 g/mol

Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 103478-63-3
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00235877
InChI Key BUJQSIPFDWLNDC-GNLPSFAGNA-N
Synonym fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl
PubChem CID 7019520
IUPAC Name (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Beilstein 7350417
Solubility Information Soluble in clear solution (0.3 gram in 2 ml DMF).
Quantity 1 g
Formula Weight 367.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Fmoc-N-methyl-D-leucine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only