missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-tyrosine, 97%

Catalog No. AAH5996703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5996703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5996703 Supplier Thermo Scientific Chemicals Supplier No. H5996703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92954-90-0 | C24H21NO5 | 403.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92954-90-0
Molecular Formula C24H21NO5
Molecular Weight (g/mol) 403.43
MDL Number MFCD00134890
InChI Key SWZCTMTWRHEBIN-ANBDAQEENA-N
Synonym fmoc-tyr-oh, fmoc-l-tyrosine, fmoc-tyrosine, nalpha-fmoc-l-tyrosine, n-fmoc-l-tyrosine, fmoc-tyr, n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosine, n, a-fmoc-l-tyrosine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, tyrosine, n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 6957986
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 182°C to 187°C
Quantity 1 g
Beilstein 3573123
Optical Rotation −20° (c=1 in DMF)
Formula Weight 403.43
Percent Purity 97%
Chemical Name or Material N-Fmoc-L-tyrosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only