missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-serine, 97+%

Catalog No. p-7025546
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2107903 1 g
AAB2107906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2107903 Supplier Thermo Scientific Chemicals Supplier No. B2107903
Only null left

CAS: 73724-45-5 | C18H16NO5 | 326.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73724-45-5
Molecular Formula C18H16NO5
Molecular Weight (g/mol) 326.33
MDL Number MFCD00051928
InChI Key JZTKZVJMSCONAK-INIZCTEOSA-M
Synonym fmoc-ser-oh, fmoc-l-serine, fmoc-l-ser-oh, n-fmoc-l-serine, n-9-fluorenylmethoxycarbonyl-l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-l-serine, n-9h-fluoren-9-ylmethoxycarbonyl serine, s-fmoc-2-amino-3-hydroxypropionic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-serine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid
PubChem CID 6541433
SMILES OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Melting Point 102°C to 106°C
Quantity 1 g
Beilstein 4715791
Optical Rotation +13.6° (c=1 in Ethyl Acetate)
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
Formula Weight 327.34
Percent Purity ≥97%
Chemical Name or Material N-Fmoc-L-serine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only