missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-methionine sulfoxide, 98%

CAS: 76265-70-8 | C20H21NO5S | 387.45 g/mol

$67.79 - $217.14

Chemical Identifiers

CAS 76265-70-8
Molecular Formula C20H21NO5S
Molecular Weight (g/mol) 387.45
MDL Number MFCD00077064
InChI Key CEHRSUBRZOGRSW-HSYKDVHTSA-N
Synonym fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
PubChem CID 7408215
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6327103
View Documents
Thermo Scientific Chemicals
H6327103
1 g
Each for $67.79
Only null left
 
AAH6327106
View Documents
Thermo Scientific Chemicals
H6327106
5 g
Each for $217.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

76265-70-8
387.45
CEHRSUBRZOGRSW-HSYKDVHTSA-N
7408215
CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C20H21NO5S
MFCD00077064
fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid

Specifications

76265-70-8
C20H21NO5S
fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1 g
7408215
98%
N-Fmoc-L-methionine sulfoxide
White
MFCD00077064
CEHRSUBRZOGRSW-HSYKDVHTSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid
387.45
387.45
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only