missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-methionine sulfoxide, 98%

Catalog No. AAH6327103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6327103 1 g
AAH6327106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6327103 Supplier Thermo Scientific Chemicals Supplier No. H6327103
Only null left

CAS: 76265-70-8 | C20H21NO5S | 387.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76265-70-8
Molecular Formula C20H21NO5S
Molecular Weight (g/mol) 387.45
MDL Number MFCD00077064
InChI Key CEHRSUBRZOGRSW-HSYKDVHTSA-N
Synonym fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
PubChem CID 7408215
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 387.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-methionine sulfoxide
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only