missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-homoleucine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6227003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Fmoc-L-homoleucine | |
| C22H25NO4 | |
| fmoc-holeu-oh, fmoc-l-homoleucine, fmoc-hle-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-methylhexanoic acid, fmoc-homoleu-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homoleucine, fmoc-s-2-amino-5-methylhexanoic acid, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, fmoc-l-hol | |
| CC(C)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 1 g | |
| 17040135 | |
| 95% |
| 180414-94-2 | |
| MFCD00270206 | |
| UJOQOPBFLFQOJJ-GNLPSFAGNA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid | |
| 367.45 | |
| 367.44 | |
| Solid |
Chemical Identifiers
| 180414-94-2 | |
| 367.45 | |
| UJOQOPBFLFQOJJ-GNLPSFAGNA-N | |
| 17040135 | |
| CC(C)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C22H25NO4 | |
| MFCD00270206 | |
| fmoc-holeu-oh, fmoc-l-homoleucine, fmoc-hle-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-methylhexanoic acid, fmoc-homoleu-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homoleucine, fmoc-s-2-amino-5-methylhexanoic acid, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, fmoc-l-hol | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only