missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-homoleucine, 95%, Thermo Scientific Chemicals

Catalog No. AAH62270MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62270MD 250 mg
AAH6227003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62270MD Supplier Thermo Scientific Chemicals Supplier No. H62270MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 180414-94-2
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00270206
InChI Key UJOQOPBFLFQOJJ-GNLPSFAGNA-N
Synonym fmoc-holeu-oh, fmoc-l-homoleucine, fmoc-hle-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-methylhexanoic acid, fmoc-homoleu-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homoleucine, fmoc-s-2-amino-5-methylhexanoic acid, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, fmoc-l-hol
PubChem CID 17040135
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid
SMILES CC(C)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 367.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-homoleucine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only