missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid, 95%

Catalog No. AAH6293622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6293622 100 g
AAH6293606 5 g
AAH6293614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6293622 Supplier Thermo Scientific Chemicals Supplier No. H6293622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 121343-82-6 | C20H19NO6 | 369.373 g/mol

N-Fmoc-L-glutamic acid nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-glutamic acid nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 121343-82-6
Molecular Formula C20H19NO6
Molecular Weight (g/mol) 369.373
MDL Number MFCD00237657
InChI Key QEPWHIXHJNNGLU-KRWDZBQOSA-N
Synonym fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid
PubChem CID 7019018
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Quantity 100 g
Beilstein 3633624
Solubility Information Slightly soluble in water.
Formula Weight 369.37
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-glutamic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only