missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%

Catalog No. p-7039765
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5916906 5 g
AAH5916914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5916906 Supplier Thermo Scientific Chemicals Supplier No. H5916906
Only null left

CAS: 71989-18-9 | C24H27NO6 | 425.481 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71989-18-9
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.481
MDL Number MFCD00037135
InChI Key OTKXCALUHMPIGM-FQEVSTJZSA-N
Synonym fmoc-glu otbu-oh, fmoc-l-glutamic acid 5-tert-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-fmoc-l-glutamic acid 5-tert-butyl ester, fmoc-glu obut, fmoc-glu tbu, fmoc-l-glu otbu, pubchem12950, fmoc-l-glu otbu-oh
PubChem CID 2724637
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 83°C to 90°C
Quantity 5 g
Beilstein 3636375
Optical Rotation −5.0° (c=1 in Acetic Acid/Water (4:1))
Formula Weight 425.48
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-glutamic acid 5-tert-butyl ester
EINECSNumber 276-253-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only