missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-methyl ester, 97%

CAS: 145038-50-2 | C21H21NO6 | 383.40 g/mol

$92.97 - $320.34

Chemical Identifiers

CAS 145038-50-2
Molecular Formula C21H21NO6
Molecular Weight (g/mol) 383.40
MDL Number MFCD01076132
InChI Key RULINAWEYRMHHQ-UHFFFAOYNA-N
Synonym fmoc-glu ome-oh, n-fmoc-l-glutamic acid 5-methyl ester, fmoc-l-glu me-oh, fmoc-l-glu me-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-fmoc-l-glutamic acid alpha-methyl ester, pubchem13156, pubchem13156, a-methyl ester, a-methyl ester
PubChem CID 51340556
SMILES COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6335806
View Documents
Thermo Scientific Chemicals
H6335806
5 g
Each for $92.97
Only null left
 
AAH6335814
View Documents
Thermo Scientific Chemicals
H6335814
25 g
Each for $320.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

145038-50-2
383.40
RULINAWEYRMHHQ-UHFFFAOYNA-N
51340556
C21H21NO6
MFCD01076132
fmoc-glu ome-oh, n-fmoc-l-glutamic acid 5-methyl ester, fmoc-l-glu me-oh, fmoc-l-glu me-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-fmoc-l-glutamic acid alpha-methyl ester, pubchem13156, pubchem13156, a-methyl ester, a-methyl ester
COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

145038-50-2
C21H21NO6
fmoc-glu ome-oh, n-fmoc-l-glutamic acid 5-methyl ester, fmoc-l-glutamic acid gamma-methyl ester, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methoxy-5-oxopentanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, ambotzfaa1720, pubchem13156, pubchem13156, fmoc-l-glutamic acid, a-methyl ester
COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
5 g
51340556
97%
N-Fmoc-L-glutamic acid 5-methyl ester
White
MFCD01076132
RULINAWEYRMHHQ-UHFFFAOYNA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methoxy-5-oxopentanoic acid
383.40
383.39
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only