missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-methyl ester, 97%

Catalog No. AAH6335806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6335806 5 g
AAH6335814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6335806 Supplier Thermo Scientific Chemicals Supplier No. H6335806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 145038-50-2 | C21H21NO6 | 383.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145038-50-2
Molecular Formula C21H21NO6
Molecular Weight (g/mol) 383.40
MDL Number MFCD01076132
InChI Key RULINAWEYRMHHQ-UHFFFAOYNA-N
Synonym fmoc-glu ome-oh, fmoc-l-glutamic acid gamma-methyl ester, fmoc-l-glu me-oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methoxy-5-oxopentanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-fmoc-l-glutamic acid alpha-methyl ester, ambotzfaa1720, pubchem13156, fmoc-l-glutamic acid, a-methyl ester, n-fmoc-l-glutamic acid 5-methyl ester
PubChem CID 51340556
SMILES COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methoxy-5-oxopentanoic acid
Quantity 5 g
Formula Weight 383.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 5-methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only