missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-allyl ester, 95%

CAS: 133464-46-7 | C23H23NO6 | 409.44 g/mol

$164.16 - $708.58

Chemical Identifiers

CAS 133464-46-7
Molecular Formula C23H23NO6
Molecular Weight (g/mol) 409.44
MDL Number MFCD00190879
InChI Key LRBARFFNYOKIAX-UHFFFAOYNA-N
Synonym fmoc-glu oall-oh, fmoc-l-glutamic acid 5-allyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, ambotzfaa1372, 5-allyl n-fmoc-l-glutamate, n-fmoc-l-glutamic acid 5-allyl ester, fmoc-glu oall-oh hplc, 5-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 5-allyl ester, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-allyl ester
PubChem CID 7019067
SMILES OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6638506
View Documents
Thermo Scientific Chemicals
H6638506
5 g
Each for $164.16
Only null left
 
AAH6638514
View Documents
Thermo Scientific Chemicals
H6638514
25 g
Each for $708.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

133464-46-7
409.44
LRBARFFNYOKIAX-UHFFFAOYNA-N
7019067
C23H23NO6
MFCD00190879
fmoc-glu oall-oh, fmoc-l-glutamic acid 5-allyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, ambotzfaa1372, 5-allyl n-fmoc-l-glutamate, n-fmoc-l-glutamic acid 5-allyl ester, fmoc-glu oall-oh hplc, 5-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 5-allyl ester, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-allyl ester
OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

133464-46-7
C23H23NO6
6670846
LRBARFFNYOKIAX-UHFFFAOYNA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-prop-2-enoxypentanoic acid
409.44
409.44
Powder
White
MFCD00190879
fmoc-glu oall-oh, fmoc-l-glutamic acid 5-allyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, ambotzfaa1372, 5-allyl n-fmoc-l-glutamate, n-fmoc-l-glutamic acid 5-allyl ester, fmoc-glu oall-oh hplc, 5-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 5-allyl ester, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-allyl ester
OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
5 g
7019067
95%
N-Fmoc-L-glutamic acid 5-allyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only