missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid, 95%

CAS: 119062-05-4 | C19H17NO6 | 355.346 g/mol

$78.19 - $321.29

Chemical Identifiers

CAS 119062-05-4
Molecular Formula C19H17NO6
Molecular Weight (g/mol) 355.346
MDL Number MFCD00237654
InChI Key KSDTXRUIZMTBNV-INIZCTEOSA-N
Synonym fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid
PubChem CID 7019016
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6227406
View Documents
Thermo Scientific Chemicals
H6227406
5 g
Each for $78.19
Only null left
 
AAH6227414
View Documents
Thermo Scientific Chemicals
H6227414
25 g
Each for $321.29
Only null left
 
Description

Description

N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

Solubility
Soluble in methanol, and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

119062-05-4
355.346
KSDTXRUIZMTBNV-INIZCTEOSA-N
7019016
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O
C19H17NO6
MFCD00237654
fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid

Specifications

119062-05-4
MFCD00237654
3632557
Soluble in methanol,and water.
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O
355.346
355.34
N-Fmoc-L-aspartic acid
C19H17NO6
5 g
fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid
KSDTXRUIZMTBNV-INIZCTEOSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid
7019016
95%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only